Skip to Content
Merck
All Photos(1)

Key Documents

200042

Sigma-Aldrich

Chromyl chloride

≥99.99% trace metals basis

Synonym(s):

Chromium(VI) dichloride dioxide, Chromium(VI) oxychloride, Dichlorochromium(VI) oxide

Sign Into View Organizational & Contract Pricing

Select a Size

10 G
791,00 kr.

791,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
10 G
791,00 kr.

About This Item

Linear Formula:
CrO2Cl2
CAS Number:
Molecular Weight:
154.90
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
for analytical purposes
form:
liquid

791,00 kr.


Please contact Customer Service for Availability

Request a Bulk Order

grade

for analytical purposes

Quality Level

Assay

≥99.99% trace metals basis

form

liquid

impurities

≤100.0 ppm Trace Metal Analysis

bp

117 °C (lit.)

mp

−96.5 °C (lit.)

density

1.911 g/mL at 25 °C (lit.)

SMILES string

Cl[Cr](Cl)(=O)=O

InChI

1S/2ClH.Cr.2O/h2*1H;;;/q;;+2;;/p-2

InChI key

AHXGRMIPHCAXFP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 1B - Eye Dam. 1 - Muta. 1B - Ox. Liq. 1 - Skin Corr. 1A - Skin Sens. 1

Storage Class Code

5.1A - Strongly oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service