Skip to Content
Merck
All Photos(1)

Key Documents

195391

Sigma-Aldrich

3-[3-(Trifluoromethyl)phenoxy]benzaldehyde

97%

Synonym(s):

3-(α,α,α-Trifluoro-m-tolyloxy)benzaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3C6H4OC6H4CHO
CAS Number:
Molecular Weight:
266.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Pricing and availability is not currently available.

Assay

97%

refractive index

n20/D 1.538 (lit.)

bp

130 °C/1.4 mmHg (lit.)

density

1.284 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)c1cccc(Oc2cccc(C=O)c2)c1

InChI

1S/C14H9F3O2/c15-14(16,17)11-4-2-6-13(8-11)19-12-5-1-3-10(7-12)9-18/h1-9H

InChI key

RPORRWJOHNQOHN-UHFFFAOYSA-N

Application

3-[3-(Trifluoromethyl)phenoxy]benzaldehyde was used in the synthesis of:
  • 5,10,15,20-tetra[3-(3-trifluoromethyl)phenoxy]porphyrin[1]
  • 4-{4-naphthalen-2-yl-5-[3-(3-trifluoromethyl-phenoxy)-phenyl]-imidazol-1-yl}-piperidine[2]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Parallel synthesis of arrays of 1, 4, 5-trisubstituted 1-(4-piperidyl)-imidazoles by IMCR: a novel class of aspartyl protease inhibitors.
Domling A, et al.
ARKIVOC (Gainesville, FL, United States), 12, 99-109 (2007)
Cynthia P Tidwell et al.
Molecules (Basel, Switzerland), 12(7), 1389-1398 (2007-10-03)
The newly synthesized 5,10,15,20-tetra[3-(3-trifluoromethyl)phenoxy]porphyrin, TTFMPP, has been characterized using mass spectroscopy, 1H-, 13C- and 19F-NMR, MALDI-TOF mass spectrometry, UV-Vis and fluorescence spectrophotometry, and cyclic voltammetry. The NMR confirmed the structure of the compound and the mass spectrum was in agreement

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service