Skip to Content
Merck
All Photos(2)

Documents

193585

Sigma-Aldrich

2,6-Dichloro-3-nitropyridine

technical grade, 92%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H2Cl2N2O2
CAS Number:
Molecular Weight:
192.99
Beilstein:
1619741
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39151701
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Assay

92%

mp

55-60 °C (lit.)

SMILES string

[O-][N+](=O)c1ccc(Cl)nc1Cl

InChI

1S/C5H2Cl2N2O2/c6-4-2-1-3(9(10)11)5(7)8-4/h1-2H

InChI key

SHCWQWRTKPNTEM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2,6-Dichloro-3-nitropyridine undergoes macrocyclic condensation reaction with resorcinol derivatives to yield chiral tetraoxacalix[2]arene[2]pyridines.

Application

2,6-Dichloro-3-nitropyridine was used:
  • in the synthesis of pyridyldifluoroacetates
  • as starting reagent in the preparation of bicyclooxacalixhetarene

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wouter Maes et al.
Chemical Society reviews, 37(11), 2393-2402 (2008-10-25)
Oxacalix[n]arenes, reassessed members of the calixarene family in which the traditional methylene bridges are replaced by oxygen atoms, have emerged as a promising class of macrocycles in recent years. This tutorial review summarizes the synthetic progress made in the field
Shuai Pan et al.
Organic letters, 14(24), 6254-6257 (2012-12-12)
Inherently chiral tetraoxacalix[2]arene[2]pyridines containing C(2) symmetry were synthesized efficiently from a macrocyclic condensation reaction of resorcinol derivatives with 2,6-dichloro-3-nitropyridine in a one-pot reaction manner, while tetraoxacalix[2]arene[2]pyridine with an ABCD-substitution pattern was prepared in a good yield by means of a
Copper-mediated reaction of 2-halopyridines with ethyl bromodifluoroacetate.
Ashwood MS, et al.
Tetrahedron Letters, 43(50), 9271-9273 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service