Skip to Content
Merck
All Photos(1)

Documents

190446

Sigma-Aldrich

Benzotriazole

reagent grade, 97%

Synonym(s):

1,2,3-Benzotriazole, 1H-Benzotriazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H5N3
CAS Number:
Molecular Weight:
119.12
Beilstein:
112133
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

reagent grade

vapor density

4.1 (vs air)

vapor pressure

0.04 mmHg ( 20 °C)

Assay

97%

mp

97-99 °C (lit.)

SMILES string

c1ccc2[nH]nnc2c1

InChI

1S/C6H5N3/c1-2-4-6-5(3-1)7-9-8-6/h1-4H,(H,7,8,9)

InChI key

QRUDEWIWKLJBPS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

338.0 °F - closed cup

Flash Point(C)

170 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Qiang Huang et al.
European journal of medicinal chemistry, 46(11), 5680-5687 (2011-09-20)
Previously, we have reported the design and synthesis of 4-aryl-1H-1,2,3-triazoles as inhibitors of indoleamine 2,3-dioxygenase (IDO), a promising therapeutic target of cancer. Here, we present the structure-activity relationship and enzyme kinetic studies on a series of 4-aryl-1H-1,2,3-triazoles. Three compounds (1
Rozanna Avagyan et al.
Environmental science and pollution research international, 22(8), 5842-5849 (2014-10-25)
Textiles play an important role in our daily life, and textile production is one of the oldest industries. In the manufacturing chain from natural and/or synthetic fibers to the final clothing products, the use of many different chemicals is ubiquitous.
K Pavić et al.
European journal of medicinal chemistry, 86, 502-514 (2014-09-10)
Novel primaquine semicarbazides 7a-l and ureas 9a-g with modified benzhydryl, trityl, phenyl or hydroxyalkyl substituents were prepared and evaluated for cytostatic and antioxidative activities. Two synthetic approaches for preparation of the title semicarbazides were applied, both having certain advantages. In
Evangelia Terzopoulou et al.
Environmental science and pollution research international, 22(2), 1095-1112 (2014-08-12)
A multi-residue method, based on gas chromatography coupled to tandem mass spectrometry (GC-MS/MS), has been developed for the determination of 70 organic micropollutants from various chemical classes (organochlorinated, organophosphorous, triazines, carbamate and urea, polycyclic aromatic hydrocarbons, polychlorinated biphenyls, pharmaceuticals, phenols
Paul Thellmann et al.
Environmental science and pollution research international, 22(21), 16405-16416 (2014-11-14)
In order to evaluate surface water and the sediment quality of rivers connected to wastewater treatment plants (WWTPs) with different treatment technologies, fish embryo tests (FET) with Danio rerio were conducted using native water and sediment samples collected upstream and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service