Skip to Content
Merck
All Photos(1)

Key Documents

178365

Sigma-Aldrich

1,4-Dioxa-8-azaspiro[4.5]decane

98%

Synonym(s):

4-Piperidone ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18
Beilstein:
506799
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
liquid
Assay:
98%

Assay

98%

form

liquid

refractive index

n20/D 1.4819 (lit.)

bp

108-111 °C/26 mmHg (lit.)

density

1.117 g/mL at 20 °C (lit.)

functional group

ether
ketal

SMILES string

C1CC2(CCN1)OCCO2

InChI

1S/C7H13NO2/c1-3-8-4-2-7(1)9-5-6-10-7/h8H,1-6H2

InChI key

KPKNTUUIEVXMOH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Dioxa-8-azaspiro[4.5]decane was used in the synthesis of 1,4-dioxa-8-azaspiro [4,5] deca spirocyclotriphosphazenes[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

177.8 °F - closed cup

Flash Point(C)

81 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nuran Asmafiliz et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 86, 214-223 (2011-11-23)
The Schiff base compounds (1 and 2) are synthesized by the condensation reactions of 2-furan-2-yl-methylamine with 2-hydroxy-3-methoxy- and 2-hydroxy-5-methoxy-benzaldehydes and reduced with NaBH(4) to give the new N/O-donor-type ligands (3 and 4). The monospirocyclotriphosphazenes containing 1,3,2-oxazaphosphorine rings (5 and 6)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service