Skip to Content
Merck
All Photos(1)

Key Documents

17816

Sigma-Aldrich

N-Fmoc-Freidinger′s lactam

≥95.0% (HPLC)

Synonym(s):

Fmoc-Gly-Val Freidinger’s lactam

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H26N2O5
CAS Number:
Molecular Weight:
422.47
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Quality Level

Assay

≥95.0% (HPLC)

form

powder or crystals

color

white

application(s)

peptide synthesis

functional group

Fmoc

storage temp.

2-8°C

SMILES string

CC(C)[C@H](N1CC[C@@H](NC(=O)OCC2c3ccccc3-c4ccccc24)C1=O)C(O)=O

InChI

1S/C24H26N2O5/c1-14(2)21(23(28)29)26-12-11-20(22(26)27)25-24(30)31-13-19-17-9-5-3-7-15(17)16-8-4-6-10-18(16)19/h3-10,14,19-21H,11-13H2,1-2H3,(H,25,30)(H,28,29)/t20-,21+/m1/s1

InChI key

PEMHOAIHPQJNIM-RTWAWAEBSA-N

Other Notes

Fmoc-protected conformationally restricted dipeptide peptidomimetic for Gly-Val-OH.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J.A. Robl et al.
Journal of the American Chemical Society, 116, 2348-2348 (1994)
R M Freidinger et al.
Science (New York, N.Y.), 210(4470), 656-658 (1980-11-07)
An analog of luteinizing hormone-releasing hormone containing a gamma-lactam as a conformational constraint has been prepared with the use of a novel cyclization of a methionine sulfonium salt. The analog is more active as a luteinizing hormone-releasing hormone agonist that

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service