177253
Guanidine hydrochloride
99%
Synonym(s):
Aminoformamidine hydrochloride, Aminomethanamidine hydrochloride, Guanidinium chloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NH2C(=NH)NH2 · HCl
CAS Number:
Molecular Weight:
95.53
Beilstein:
3591990
EC Number:
MDL number:
UNSPSC Code:
12352200
Recommended Products
Assay
99%
mp
180-185 °C (lit.)
density
1.3 g/cm3 (lit.)
SMILES string
Cl[H].NC(N)=N
InChI
1S/CH5N3.ClH/c2-1(3)4;/h(H5,2,3,4);1H
InChI key
PJJJBBJSCAKJQF-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
Strong chaotropic agent useful for the denaturation and subsequent refolding of proteins. This strong denaturant can solubilize insoluble or denatured proteins such as inclusion bodies. This can be used as the first step in refolding proteins or enzymes into their active form. Urea and dithiothreitol (DTT) may also be necessary.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Armando M De Palma et al.
Antimicrobial agents and chemotherapy, 53(5), 1850-1857 (2009-02-25)
A novel compound, TTP-8307, was identified as a potent inhibitor of the replication of several rhino- and enteroviruses. TTP-8307 inhibits viral RNA synthesis in a dose-dependent manner, without affecting polyprotein synthesis and/or processing. Drug-resistant variants of coxsackievirus B3 were all
Caitriona McKeever et al.
Journal of medicinal chemistry, 56(3), 700-711 (2013-01-11)
Considering the strong DNA minor groove binding observed for our previous series of diaromatic symmetric and asymmetric guanidinium and 2-aminoimidazolinium derivatives, we report now the synthesis of new aminoalkyl derivatives of diaromatic guanidines with potential as DNA minor groove binders
Masaatsu Adachi et al.
The Journal of organic chemistry, 78(4), 1699-1705 (2013-01-18)
We describe an improved synthesis of (-)-5,11-dideoxytetrodotoxin from an enone, which was used for synthesis of tetrodotoxin and its analogues in this laboratory. One of the major modifications was to establish a two-step guanidinylation of trichloroacetamide of a highly functionalized
Fackson Mwale et al.
Tissue engineering. Part A, 20(21-22), 2942-2949 (2014-05-03)
Link N is a naturally occurring peptide that can stimulate proteoglycan synthesis in intervertebral disc (IVD) cells. IVD repair can also potentially be enhanced by mesenchymal stem cell (MSC) supplementation to maximize extracellular matrix (ECM) production. In a previous study
Stefan J Kempf et al.
PloS one, 9(10), e110464-e110464 (2014-10-21)
Patients suffering from brain malignancies are treated with high-dose ionising radiation. However, this may lead to severe learning and memory impairment. Preventive treatments to minimise these side effects have not been possible due to the lack of knowledge of the
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service