Skip to Content
Merck
All Photos(1)

Documents

165360

Sigma-Aldrich

2,4-Dihydroxy-5,6-dimethylpyrimidine

97%

Synonym(s):

5,6-Dimethyluracil

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H8N2O2
CAS Number:
Molecular Weight:
140.14
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

297-300 °C (dec.) (lit.)

SMILES string

CC1=C(C)C(=O)NC(=O)N1

InChI

1S/C6H8N2O2/c1-3-4(2)7-6(10)8-5(3)9/h1-2H3,(H2,7,8,9,10)

InChI key

PZVLJGKJIMBYNP-UHFFFAOYSA-N

Application

2,4-Dihydroxy-5,6-dimethylpyrimidine was used in the synthesis of mono and polyatomic titanium derivatives. It was used as staring reagent in the synthesis of 2-(4-fluorobenzoyl) substituted pyrimidine derivatives.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A family of titanium (IV) alkoxo complexes with N, O and O, O chelating ligands. Crystal structure of [Ti (O-i-Pr)2{2-(-)-menthoxo-pyridine}2].
Perez Y, et al.
Inorgorganica Chimica Acta, 360(2), 607-618 (2007)
Young Ae Yoon et al.
Bioorganic & medicinal chemistry letters, 20(19), 5735-5738 (2010-09-03)
A series of pyrimidine derivatives as acid pump antagonists (APAs) was synthesized and the inhibitory activities against H(+)/K(+) ATPase isolated from hog gastric mucosa were determined. After elaborating on substituents at C2 and C4 position of the pyrimidine scaffold, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service