Recommended Products
Assay
97%
form
solid
bp
127-130 °C/18 mmHg (lit.)
mp
73-75 °C (lit.)
SMILES string
Cc1cnc(O[Si](C)(C)C)nc1O[Si](C)(C)C
InChI
1S/C11H22N2O2Si2/c1-9-8-12-11(15-17(5,6)7)13-10(9)14-16(2,3)4/h8H,1-7H3
InChI key
DRNUNECHVNIYHQ-UHFFFAOYSA-N
Application
O,O′-Bis(trimethylsilyl)thymine is a reactive intermediate for the preparation of nucleosides.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
EJNMMI research, 3(1), 52-52 (2013-07-16)
Automated radiosynthesizers are vital for routine production of positron-emission tomography tracers to minimize radiation exposure to operators and to ensure reproducible synthesis yields. The recent trend in the synthesizer industry towards the use of disposable kits aims to simplify setup
Angewandte Chemie (International Edition in English), 8, 976-976 (1969)
A general synthesis of pyrimidine nucleosides.
Angewandte Chemie (International ed. in English), 9(6), 461-462 (1970-06-01)
A NEW SYNTHETIC METHOD OF NUCLEOSIDES.
Chemical & pharmaceutical bulletin, 11, 1470-1472 (1963-11-01)
Studies on synthetic nucleosides. I. Trimethylsilyl derivatives of pyrimidines and purines.
Chemical & pharmaceutical bulletin, 12(3), 352-356 (1964-03-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service