Skip to Content
Merck
All Photos(3)

Documents

15567

Sigma-Aldrich

N-Boc-N-methylethylenediamine

≥97.0% (GC)

Synonym(s):

tert-Butyl N-(2-aminoethyl)-N-methylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H18N2O2
CAS Number:
Molecular Weight:
174.24
Beilstein:
3588326
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.22

Assay

≥97.0% (GC)

form

liquid

reaction suitability

reagent type: cross-linking reagent

refractive index

n20/D 1.447

density

0.975 g/mL at 20 °C (lit.)

functional group

Boc
amine

SMILES string

NCCN(C)C(OC(C)(C)C)=O

InChI

1S/C8H18N2O2/c1-8(2,3)12-7(11)10(4)6-5-9/h5-6,9H2,1-4H3

InChI key

QYJVBVKFXDHFPQ-UHFFFAOYSA-N

Other Notes

Mono-protected building block

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

>212.0 °F - closed cup

Flash Point(C)

> 100.0 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

J.A.W. Kruijtzer et al.
Tetrahedron Letters, 38, 5335-5335 (1997)
G. Eisenbrand et al.
Synthesis, 1247-1247 (1996)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service