Skip to Content
Merck
All Photos(1)

Key Documents

153168

Sigma-Aldrich

p-Chlorophenoxyacetic acid

98%

Synonym(s):

4-CPA, 4-Chlorophenoxyacetic acid, CPA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H4OCH2CO2H
CAS Number:
Molecular Weight:
186.59
Beilstein:
1211804
EC Number:
MDL number:
UNSPSC Code:
12352100

Assay

98%

mp

157-159 °C (lit.)

SMILES string

OC(=O)COc1ccc(Cl)cc1

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sonia Traverso et al.
The Journal of general physiology, 122(3), 295-306 (2003-08-13)
Opening of CLC chloride channels is coupled to the translocation of the permeant anion. From the recent structure determination of bacterial CLC proteins in the closed and open configuration, a glutamate residue was hypothesized to form part of the Cl--sensitive
B Kaya et al.
Teratogenesis, carcinogenesis, and mutagenesis, 19(4), 305-312 (1999-07-16)
The phenoxyacetates 2,4-D and 4-CPA were evaluated for genotoxicity using the Drosophila melanogaster wing spot test, which assesses for somatic mutation and recombination events. Third-instar larvae trans-heterozygous for two recessive mutations affecting the expression of wing trichomes, multiple wing hairs
Xia Li
Guang pu xue yu guang pu fen xi = Guang pu, 22(1), 57-58 (2003-08-28)
This work has synthesized a title complex using rare earth ion Eu3+ with the o-chlorophenoxyacetic acid and 2,2'-bipyridine. The chemical formula of the complex was determined to be Eu(o-ClC6H4OCH2COO)3.2,2'-bipy by elemental analysis, analysis of IR, UV, fluorescence spectra. The ternary
Imre Zs-Nagy
Annals of the New York Academy of Sciences, 959, 308-320 (2002-04-27)
As was shown in a recent review by this author (Ann. N.Y. Acad. Sci., 928: 187-199, 2001), oxyradicals cannot be considered only as harmful by-products of the oxidative metabolism, but living cells and organisms implicitly require their production. This idea
Y Alicigüzel et al.
Redox report : communications in free radical research, 6(3), 153-154 (2001-08-29)
To investigate the possible role of oxygen free radicals and oxidant stress in the toxic effects of phenoxyherbicides, we studied the in vitro effect of 4-chlorophenoxyacetic acid (4-CPA) on various human erythrocyte antioxidant enzymes, namely glucose-6-phosphate dehydrogenase, catalase, selenium-dependent glutathione

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service