Skip to Content
Merck
All Photos(1)

Key Documents

151343

Sigma-Aldrich

2-Methoxy-1-naphthaldehyde

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC10H6CHO
CAS Number:
Molecular Weight:
186.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

form

solid

bp

205 °C/18 mmHg (lit.)

mp

82-85 °C (lit.)

SMILES string

COc1ccc2ccccc2c1C=O

InChI

1S/C12H10O2/c1-14-12-7-6-9-4-2-3-5-10(9)11(12)8-13/h2-8H,1H3

InChI key

YIQGLTKAOHRZOL-UHFFFAOYSA-N

General description

The FT-IR and FT-Raman spectra of 2-Methoxy-1-naphthaldehyde was studied.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective fluorescent labeling of amino groups of bovine pancreatic trypsin inhibitor by reductive alkylation.
D Amir et al.
Biopolymers, 25(9), 1645-1658 (1986-09-01)
V Krishnakumar et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 63(2), 464-476 (2005-07-02)
FT-IR and FT-Raman spectra of 2-hydroxy-3-methoxy-5-nitrobenzaldehyde (HMN) and 2-methoxy-1-naphthaldehyde (MN) have been recorded in the regions 4000-400 cm(-1) and 3500-100 cm(-1), respectively. The molecular structure, conformational stability, geometry optimization, vibrational frequencies have been investigated. The total energy calculations of HMN

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service