Skip to Content
Merck
All Photos(1)

Key Documents

15006

Sigma-Aldrich

Boc-Phe(4-Me)-OH

≥98.0% (TLC)

Synonym(s):

Boc-4-methyl-L-phenylalanine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H21NO4
CAS Number:
Molecular Weight:
279.33
Beilstein:
5381557
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

Assay

≥98.0% (TLC)

optical activity

[α]20/D +16±2°, c = 1% in methanol

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

84-88 °C

application(s)

peptide synthesis

SMILES string

Cc1ccc(C[C@H](NC(=O)OC(C)(C)C)C(O)=O)cc1

InChI

1S/C15H21NO4/c1-10-5-7-11(8-6-10)9-12(13(17)18)16-14(19)20-15(2,3)4/h5-8,12H,9H2,1-4H3,(H,16,19)(H,17,18)/t12-/m0/s1

InChI key

JYRWNPUFECDJCX-LBPRGKRZSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gergana Shipkovenska et al.
eLife, 9 (2020-06-04)
Heterochromatic domains containing histone H3 lysine 9 methylation (H3K9me) can be epigenetically inherited independently of underlying DNA sequence. To gain insight into the mechanisms that mediate epigenetic inheritance, we used a Schizosaccharomyces pombe inducible heterochromatin formation system to perform a
Justyna Sawa-Makarska et al.
Science (New York, N.Y.), 369(6508) (2020-09-05)
Autophagosomes form de novo in a manner that is incompletely understood. Particularly enigmatic are autophagy-related protein 9 (Atg9)-containing vesicles that are required for autophagy machinery assembly but do not supply the bulk of the autophagosomal membrane. In this study, we
Saya Ito et al.
Biochimica et biophysica acta. Gene regulatory mechanisms (2018-08-05)
The androgen receptor (AR) promotes growth of prostate cancer cells by controlling the expression of target genes. This study showed that MRG domain binding protein (MRGBP) accelerated AR-mediated transactivation. We first showed that MRGBP promoted growth of AR-positive prostate cancer
Takuya Inoue et al.
European journal of pharmacology, 858, 172492-172492 (2019-06-25)
Lipoprotein lipase (LPL) is the rate-controlling enzyme for the accumulation of triacylglycerol into adipocytes, which acts by digesting it into glycerol and fatty acids. In this study, we found that treatment with (+)-JQ1, an inhibitor of the bromodomain and extra-terminal
Jing Luan et al.
Cell reports, 34(8), 108783-108783 (2021-02-25)
CCCTC-binding factor (CTCF) is a conserved zinc finger transcription factor implicated in a wide range of functions, including genome organization, transcription activation, and elongation. To explore the basis for CTCF functional diversity, we coupled an auxin-induced degron system with precision

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service