Skip to Content
Merck
All Photos(1)

Key Documents

148954

Sigma-Aldrich

Ethyl undecanoate

97%

Synonym(s):

Undecanoic acid ethyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9CO2C2H5
CAS Number:
Molecular Weight:
214.34
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.428 (lit.)

bp

105 °C/4 mmHg (lit.)

density

0.859 g/mL at 25 °C (lit.)

functional group

ester

SMILES string

CCCCCCCCCCC(=O)OCC

InChI

1S/C13H26O2/c1-3-5-6-7-8-9-10-11-12-13(14)15-4-2/h3-12H2,1-2H3

InChI key

IAFQYUQIAOWKSB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl undecanoate was used to prepare the corresponding aldehyde in the presence of Lithium diisobutyl-t-butoxyaluminum hydride (LDBBA)[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lithium diisobutyl-t-butoxyaluminum hydride, a new and efficient reducing agent for the conversion of esters to aldehydes.
Tetrahedron Letters, 48(29), 5061-5064 (2007)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service