Skip to Content
Merck
All Photos(1)

Key Documents

148121

Sigma-Aldrich

3,4-Dinitrotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)2
CAS Number:
Molecular Weight:
182.13
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

mp

54-57 °C (lit.)

SMILES string

Cc1ccc(c(c1)[N+]([O-])=O)[N+]([O-])=O

Application

3,4-dinitrotoluene was used as an internal standard in the GC/MS and GC/MS/MS experiments. It was also used to study the solubility of 3,4-dinitrotoluene in pure and saline water.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2 - Carc. 1B - Muta. 2 - Repr. 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R J Spanggord et al.
Chemical research in toxicology, 3(6), 551-558 (1990-11-01)
The relation of various structural parameters to hepatotoxic potential was investigated by using six dinitrotoluene (DNT) isomers and isolated rat hepatocyte suspensions as the biological test system. DNT-induced hepatotoxicity was found to correlate with an inhibition of protein synthesis and
I Holen et al.
Journal of toxicology and environmental health, 29(1), 89-98 (1990-01-01)
The effects of four isomers of dinitrotoluene (DNT) and technical DNT (a mixture of DNT isomers and other compounds, with 2,4-DNT as the major constituent) were studied in two short-term in vitro assays. None of the isomers or technical DNT
Jeannette M Perr et al.
Talanta, 67(2), 430-436 (2008-10-31)
The characteristic ions or the parent ion resulting from ionization can be isolated in an ion trap and subjected to further fragmentation during a gas chromatography-tandem mass spectrometry (GC/MS/MS) experiment. This approach can improve the selectivity and sensitivity of explosive
D R Sorenson et al.
Cell biology and toxicology, 19(2), 107-119 (2003-06-05)
In this study we test the hypothesis that immortalized adult rat Sertoli cells respond to known testicular toxins in a similar manner to Sertoli cells tested in vivo and in primary culture. This cell line was developed by immortalizing adult
Yana A Kholod et al.
Chemosphere, 83(3), 287-294 (2011-01-11)
The solubility in pure and saline water at various temperatures was calculated for selected nitro compounds (nitrobenzene, 1,3,5-trinitrobenzene, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, 2,4-dinitrotoluene, 2,6-dinitrotoluene, 2,3-dinitrotoluene, 3,4-dinitrotoluene, 2,4,6-trinitrotoluene) using the Conductor-like Screening model for Real Solvents (COSMO-RS). The results obtained were compared

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service