Skip to Content
Merck
All Photos(3)

Documents

143596

Sigma-Aldrich

2,5-Dimethoxycinnamic acid, predominantly trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3O)2C6H3CH=CHCO2H
CAS Number:
Molecular Weight:
208.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

mp

148-150 °C (lit.)

SMILES string

COc1ccc(OC)c(\C=C\C(O)=O)c1

InChI

1S/C11H12O4/c1-14-9-4-5-10(15-2)8(7-9)3-6-11(12)13/h3-7H,1-2H3,(H,12,13)/b6-3+

InChI key

JPQWWJZORKTMIZ-ZZXKWVIFSA-N

General description

2,5-Dimethoxycinnamic acid forms 1:1 molecular complex with 3,5-dinitrocinnamic acid.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Crystal Engineering-Where Do We Go from Here?
Sharma CVK.
Crystal Growth & Design, 2(6), 465-474 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service