Skip to Content
Merck
All Photos(1)

Key Documents

131512

Sigma-Aldrich

Picrolonic acid

97%

Synonym(s):

2,4-Dihydro-5-methyl-4-nitro-2-(4-nitrophenyl)-3H-pyrazol-3-one, 3-Methyl-4-nitro-1-(4-nitrophenyl)-2-pyrazolin-5-one, 3-Methyl-4-nitro-1-(p-nitrophenyl)-5-pyrazolone, Picrolic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H8N4O5
CAS Number:
Molecular Weight:
264.19
Beilstein:
36329
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:

Assay

97%

SMILES string

CC1=NN(c2ccc(cc2)[N+]([O-])=O)C(=O)C1[N+]([O-])=O

InChI

1S/C10H8N4O5/c1-6-9(14(18)19)10(15)12(11-6)7-2-4-8(5-3-7)13(16)17/h2-5,9H,1H3

InChI key

OVFUUSPKWADLNJ-UHFFFAOYSA-N


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The composition of crystalline secretin picrolonate.
H GREENGARD et al.
Federation proceedings, 6(1 Pt 2), 115-115 (1947-01-01)
P G Veltsistas et al.
The Analyst, 119(7), 1613-1617 (1994-07-01)
The construction and some analytical applications of a liquid picrolonate (Picrl-) ion-selective electrode based on the tris(phenanthrolinato)iron(II) (ferroin) picrolonate salt, dissolved in 2-nitrotoluene, are described. The liquid membrane electrode exhibits a rapid and almost Nernstian response to picrolonate anions in
H Lee et al.
Mutation research, 204(2), 229-234 (1988-02-01)
The antimutagenic activities of extracts of 36 commonly used anticancer crude drugs from Chinese herbs were studied by using the Salmonella/microsomal system in the presence of picrolonic acid or benzo[a]pyrene to test whether they contain direct or indirect antimutagens. Each
M Apostu et al.
Revista medico-chirurgicala a Societatii de Medici si Naturalisti din Iasi, 109(2), 422-425 (2006-04-13)
Famotidine, belonging to H2-antagonist group, is a compound containing a thiazolic moiety and it is used in peptic ulcer therapy. This paper debates the possibility of developing a new ultraviolet spectrophotometric assessment by using the reaction between famotidine and picrolonic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service