11927
2-Chloropropionaldehyde dimethyl acetal
produced by Wacker Chemie AG, Burghausen, Germany, ≥96.0% (GC)
Synonym(s):
Cl-PADMA, 2-Chloro-1,1-dimethoxypropane
About This Item
Recommended Products
grade
produced by Wacker Chemie AG, Burghausen, Germany
Assay
≥96.0% (GC)
manufacturer/tradename
Wacker Chemie AG
bp
126-127 °C (lit.)
density
1.059 g/mL at 20 °C (lit.)
SMILES string
COC(OC)C(C)Cl
Looking for similar products? Visit Product Comparison Guide
Other Notes
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service