Skip to Content
Merck
All Photos(2)

Key Documents

115061

Sigma-Aldrich

2-Benzoyl-5-norbornene,mixture of endo and exo

97%

Synonym(s):

5-Norbornen-2-yl phenyl ketone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H14O
CAS Number:
Molecular Weight:
198.26
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

refractive index

n20/D 1.568 (lit.)

bp

122-124 °C/2 mmHg (lit.)

density

1.115 g/mL at 25 °C (lit.)

functional group

ketone
phenyl

SMILES string

O=C(C1C[C@H]2C[C@@H]1C=C2)c3ccccc3

InChI

1S/C14H14O/c15-14(11-4-2-1-3-5-11)13-9-10-6-7-12(13)8-10/h1-7,10,12-13H,8-9H2/t10-,12+,13?/m1/s1

InChI key

RMHYEJKELLBHSV-FVBASEICSA-N

General description

Norbornenes comprise of compounds with norbornene ring moiety, and also include norbornadiene, substituted norbornenes and polycyclic norbornenes. The substituted norbornenes possess a norbornene ring moiety and at least one substituent group.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service