Skip to Content
Merck
All Photos(1)

Documents

113468

Sigma-Aldrich

2-(2-Aminoethylamino)-5-nitropyridine

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H10N4O2
CAS Number:
Molecular Weight:
182.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

mp

126-128 °C (lit.)

SMILES string

NCCNc1ccc(cn1)[N+]([O-])=O

InChI

1S/C7H10N4O2/c8-3-4-9-7-2-1-6(5-10-7)11(12)13/h1-2,5H,3-4,8H2,(H,9,10)

InChI key

ODHSPTHLPCXPTL-UHFFFAOYSA-N

Application

2-(2-Aminoethylamino)-5-nitropyridine is used in matrix preparation for the quantification of phospholipids (PLs) by MALDI-TOFMS.
2-(2-Aminoethylamino)-5-nitropyridine is used to prepare guanidine.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rosendo Estrada et al.
Journal of mass spectrometry : JMS, 39(4), 412-422 (2004-04-23)
The detection of phospholipids (PLs) by matrix-assisted laser desorption/ionization time-of-flight mass spectrometry was demonstrated nearly a decade ago. However, its use as a conventional tool for PL analysis has been hindered by ambiguities in peak assignments caused by spectral overlaps
A Convenient Solid Phase Synthesis of Disubstituted Guanidines.
Sandanayake S, et al.
QSAR & Combinatorial Science, 23(8), 655-661 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service