Skip to Content
Merck
All Photos(1)

Documents

101133

Sigma-Aldrich

Butylmalonic acid

99%

Synonym(s):

1,1-Pentanedicarboxylic acid, 2-n -Butylmalonic acid, 2-Butylpropanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)3CH(CO2H)2
CAS Number:
Molecular Weight:
160.17
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

99%

form

solid

mp

105-107 °C (lit.)

SMILES string

CCCCC(C(O)=O)C(O)=O

InChI

1S/C7H12O4/c1-2-3-4-5(6(8)9)7(10)11/h5H,2-4H2,1H3,(H,8,9)(H,10,11)

InChI key

MCRZWYDXIGCFKO-UHFFFAOYSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

R B Gregory et al.
Biochimica et biophysica acta, 1137(1), 34-38 (1992-10-06)
Isolated hepatocytes from hypothyroid, euthyroid and hyperthyroid rats have been employed to investigate the relative importance of reducing-equivalent shuttles for the transfer of hydrogen between cytoplasm and mitochondria during simultaneous ureogenesis and gluconeogenesis. In cells from hypothyroid animals, a 58%
Subhendu Dhibar et al.
Dalton transactions (Cambridge, England : 2003), 48(13), 4362-4369 (2019-03-13)
A novel Co(ii)-complex {i.e. [Co(n-BuM)(DPA)(H2O)2]·H2O} [n-BuMH2 = n-butylmalonic acid and DPA = 2,2'-dipyridylamine] was synthesized. The supramolecular feature, i.e. a metal coordinated and free guest water mediated hydrogen-bonding interaction assisted supramolecular ππ assembly, has been observed in the crystal structure
H Breitbart et al.
Biochimica et biophysica acta, 1026(1), 57-63 (1990-07-09)
Calcium uptake into filipin-treated bovine spermatozoa is completely inhibited by the uncoupler CCCP or by ruthenium red. Both Pi and mitochondrial substrates are required to obtain the maximal rate of calcium uptake into the sperm mitochondria. Bicarbonate and other anions
E Fransvea et al.
Archives of biochemistry and biophysics, 355(1), 93-100 (1998-07-02)
The impermeability of the inner membrane to protons is one of the four postulates of the chemiosmotic theory on the coupling mechanism between respiration and phosphorylation in mitochondria. However, oxygen uptake in isolated nonphosphorylating mitochondria requires that protons translocated from
R S Kaplan et al.
The Journal of biological chemistry, 260(18), 10293-10298 (1985-08-25)
The mitochondrial dicarboxylate carrier has been substantially purified from rat liver mitoplasts by extraction with Triton X-114 in the presence of cardiolipin followed by chromatography on hydroxylapatite. Upon incorporation of the hydroxylapatite eluate into phospholipid vesicles, an n-butylmalonate-sensitive malonate/malate exchange

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service