(Diethylamino)sulfur trifluoride
Synonym(s):
DAST, Diethylaminosulfur trifluoride
Linear Formula:
(C2H5)2NSF3
Products
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing