Skip to Content
Merck
All Photos(1)

Documents

FAP80467

Metals in wheat

reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

reference material

Quality Level

packaging

pkg of 50 g

manufacturer/tradename

Fera Science Ltd

application(s)

food and beverages

format

matrix material

Related Categories

Other Notes

Certified for the analytes listed below. See certificate for values and more details
Trace Elements
Aluminium (Al), Arsenic (As, total), Cadmium (Cd), Copper (Cu), Iron (Fe), Lead (Pb), Mercury (Hg, total), Nickel (Ni), Zinc (Zn)

Matrix Group: Cereals, Bread, Rice and Maize

Storage Class Code

13 - Non Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service