Skip to Content
Merck
All Photos(1)

Key Documents

11887

Supelco

Coated GC Packing

phase 4% Carbowax 20M + 0.8% KOH, matrix 60/80 Carbopack B support, bottle of 15 g

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23151817

packaging

bottle of 15 g

Quality Level

parameter

60-220 °C temp. range

technique(s)

gas chromatography (GC): suitable

matrix

60/80 Carbopack B support

matrix active group

4% Carbowax 20M + 0.8% KOH phase

column type

packed GC

compatibility

for analyte group amines

Looking for similar products? Visit Product Comparison Guide

General description

Ready-to-use coated packing for those wanting to pack their own columns.
The choice of coating material is based on the stationary phase of the GC. Gas Chromatographic (GC) column packed with Carbopack B modified by Carbowax and KOH has the best separation efficiency.

Application

GC column packed with Carbopack B, Carbowax and KOH was used for the determination of Dimethylethylamine and Dimethylethylamine-N-Oxide in Air, Plasma and Urine Samples.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Methods for the determination of dimethylethylamine and dimethylethylamine-N-oxide in air, plasma and urine samples.
Lundh TB, et al.
International Journal of Environmental Analytical Chemistry, 44 (2), 81-86 (1991)
Comparison of column packings for trace analysis of free amines by gas-liquid chromatography.
Mathiasson L and Lovkvist P.
Journal of Chromatography A, 217, 177-181 (1981)
Analytical Measurements in Aquatic Environments.
Namiesnik J and Szefer P.
Science, 73-73 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service