Key Documents
Z324930
BRAND® BLAUBRAND® bulb pipette, calibrated to deliver (TD, EX)
capacity 2.5 mL, one mark
About This Item
UNSPSC Code:
41121500
NACRES:
NB.01
material
AR-Glas®
description
total delivery Ex + 15 sec waiting time
feature
class AS
packaging
pack of 6 ea
manufacturer/tradename
BRAND 29704
capacity
2.5 mL
General description
Additional to DIN
All BLAUBRAND bulb pipettes are supplied with a batch number and an accompanying batch certificate in the original packaging.
According to DIN EN ISO 648.
Other Notes
BR29704 (India)
Legal Information
AR-Glas is a registered trademark of Schott AG
BLAUBRAND is a registered trademark of BRAND GMBH + CO KG
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Visit the Document Library
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.