Skip to Content
Merck
All Photos(1)

Documents

U3385

Sigma-Aldrich

UA62784

≥98% (HPLC)

Synonym(s):

4-[5-(4-Methoxyphenyl)-2-oxazolyl]-9H-Fluoren-9-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H15NO3
CAS Number:
Molecular Weight:
353.37
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

yellow

solubility

DMSO: >2 mg/mL

storage temp.

2-8°C

SMILES string

COc1ccc(cc1)-c2cnc(o2)-c3cccc4C(=O)c5ccccc5-c34

InChI

1S/C23H15NO3/c1-26-15-11-9-14(10-12-15)20-13-24-23(27-20)19-8-4-7-18-21(19)16-5-2-3-6-17(16)22(18)25/h2-13H,1H3

InChI key

SVICLWPFAQYZLX-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Studies have also reported that UA62784 associates with tubulin at or near the colchicine-binding sites in cells and functions as a cytotoxic, microtubule inhibitor.
UA62784 is a specific inhibitor of centromere protein E (CENPE) kinesin-like protein; mitotic inhibitor. CENP-E is a kinesin-like motor protein that localizes to the kinetochore during mitosis and is essential for bipolar spindle formation. UA62784 is a novel specific inhibitor of CENP-E, which likely binds within the motor domain of CENP-E. UA62784 causes reversible cell cycle arrest in mitosis before metaphase, which leads to apoptosis. The compound is not interacting with microtubules directly.

Preparation Note

UA62784 is soluble in DMSO at a concentration >2 mg/ml.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sergey Tcherniuk et al.
Chemistry & biology, 18(5), 631-641 (2011-05-26)
A recent screen for compounds that selectively targeted pancreatic cancer cells isolated UA62784. We found that UA62784 inhibits microtubule polymerization in vitro. UA62784 interacts with tubulin dimers ten times more potently than colchicine, vinblastine, or nocodazole. Competition experiments revealed that UA62784
Julia Würtemberger et al.
PloS one, 15(9), e0238572-e0238572 (2020-09-09)
Functional genomic screening of KRAS-driven mouse sarcomas was previously employed to identify proliferation-relevant genes. Genes identified included Ubiquitin-conjugating enzyme E2 (Ube2c), Centromere Protein E (Cenpe), Hyaluronan Synthase 2 (Has2), and CAMP Responsive Element Binding Protein 3 Like 2 (Creb3l2). This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service