TMO331826-BR
Nunc® barcoded latch rack
for 5.0 mL cryogenic tubes, plastic, 30 positions, case of 10 ea
Synonym(s):
tube rack, tube storage
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122804
NACRES:
NB.12
Recommended Products
material
plastic
packaging
case of 10 ea
manufacturer/tradename
Nunc 331826-BR
technique(s)
cryopreservation: suitable
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Store tubes for automated picking/placing, liquid handling and other high throughput applications using the Thermo Scientific Nunc Universal Latch Rack for Cryogenic Tubes.
Legal Information
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service