Skip to Content
Merck
All Photos(1)

Key Documents

SRP6509

Sigma-Aldrich

Chymotrypsin from human pancreas

≥95% (SDS-PAGE)

Synonym(s):

CLCR, CTRC, CTRC_HUMAN, Caldecrin, Chymotrypsin C, Chymotrypsin-C, ELA 4, Elastase 4, Elastase IV, Serum calcium decreasing factor

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12352204
NACRES:
NA.78

biological source

human

Assay

≥95% (SDS-PAGE)

form

lyophilized

mol wt

25 kDa

packaging

pkg of 100 μg

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... CTRC(11330)

General description

Chymotrypsin from human pancreas is a serine peptidase and has 241 amino acid residues contained in three polypeptide chains (A chain-13 residues, B chain-131 residues, and C chain-97 residues) linked by disulfide bridges. It is secreted by the pancreas as inactive chymotrypsinogen C. Molecular weight of this enzyme is found to be 25kDa. The pI is 8.75.

Biochem/physiol Actions

Chymotrypsin from human pancreas selectively hydrolyzes peptide bonds on the C-terminal side of tyrosine, phenylalanine, tryptophan and leucine. Loss-of-function mutations in this gene have been associated with increased risk of chronic pancreatitis. It has high activity toward leucyl peptide bonds. The enzyme acts as a co-activator of procarboxypeptidases A1 and A2.

Physical form

Lyophilized as a salt-free solid.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mesotrypsin Signature Mutation in a Chymotrypsin C (CTRC) Variant Associated with Chronic Pancreatitis.
Szabo A
The Journal of Biological Chemistry, 290(28), 17282-17292 (2015)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service