Skip to Content
Merck
All Photos(2)

Key Documents

SML3805

Sigma-Aldrich

SIRT6-IN-1

≥98% (HPLC)

Synonym(s):

1,2,3,4-Tetrahydro-2,4-dioxo-N-[4-(3-pyridinyloxy)phenyl]-6-quinazolinesulfonamide, 2,4-Dioxo-N-[4-(3-pyridyloxy)phenyl]-1H-quinazoline-6-sulfonamide, 2,4-dioxo-N-(4-(pyridin-3-yloxy)phenyl)-1,2,3,4-tetrahydroquinazoline-6-sulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H14N4O5S
CAS Number:
Molecular Weight:
410.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.21

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

-10 to -25°C

SMILES string

[S](=O)(=O)(Nc3ccc(cc3)Oc4cnccc4)c1cc2c([nH][c]([nH][c]2=O)=O)cc1

InChI

1S/C19H14N4O5S/c24-18-16-10-15(7-8-17(16)21-19(25)22-18)29(26,27)23-12-3-5-13(6-4-12)28-14-2-1-9-20-11-14/h1-11,23H,(H2,21,22,24,25)

Biochem/physiol Actions

SIRT6-IN-1 (2,4-dioxo-N-(4-(pyridin-3-yloxy)phenyl)−1,2,3,4-tetrahydroquinazoline-6-sulfonamide) is a cell penetrant, potent and selective inhibitor of SIRT6 (Sirtuin 6). It improves glucose tolerance in a type 2 diabetes mouse model. SIRT6-IN-1 delays experimental autoimmune encephalomyelitis disease onset in mice.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sirtuin 6 inhibition protects against glucocorticoid-induced skeletal muscle atrophy by regulating IGF/PI3K/AKT signaling
Nature Communications, 13(1), 5415-5415 (2022)
Sirt6 inhibition delays the onset of experimental autoimmune encephalomyelitis by reducing dendritic cell migration
Journal of Neuroinflammation, 17(1), 228-228 (2020)
Discovery of novel and selective SIRT6 inhibitors
European Journal of Medicinal Chemistry, 57(11):4796-804., 4796-4804 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service