Skip to Content
Merck
All Photos(2)

Key Documents

SAB3501219

Sigma-Aldrich

Anti-Avian Influenza H5N1 Hemagglutinin (NT) antibody produced in goat

affinity isolated antibody

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.43
conjugate:
unconjugated
application:
ELISA
WB
clone:
polyclonal
species reactivity:
influenza A virus (avian | subtype H5N1)
citations:
technique(s):
ELISA: suitable
western blot: suitable

biological source

goat

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

liquid

species reactivity

influenza A virus (avian | subtype H5N1)

concentration

1 mg/mL

technique(s)

ELISA: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Avian Influenza Hemagglutinin 2 Antibody: Influenza A virus is a major public health threat, killing more than 30, 000 people per year in the USA. Novel influenza virus strains caused by genetic drift and viral recombination emerge periodically to which humans have little or no immunity, resulting in devastating pandemics. Influenza A can exist in a variety of animals; however it is in birds that all subtypes can be found. These subtypes are classified based on the combination of the virus coat glycoproteins hemagglutinin (HA) and neuraminidase (NA) subtypes. During 1997, an H5N1 avian influenza virus was determined to be the cause of death in 6 of 18 infected patients in Hong Kong. There was some evidence of human to human spread of this virus, but it is thought that the transmission efficiency was fairly low. HA interacts with cell surface proteins containing oligosaccharides with terminal sialyl residues. Virus isolated from a human infected with the H5N1 strain in 1997 could bind to oligosaccharides from human as well as avian sources, indicating its species-jumping ability.

Immunogen

Avian Influenza H5N1 Hemagglutinin (NT) Antibody was raised against a synthetic peptide corresponding to 12 amino acids near the amino terminus of the Hemagglutinin protein.Efforts were made to use relatively conserved regions of the viral sequence as the antigen.The immunogen is located within amino acids 110 - 160 of Avian Influenza H5N1 Hemagglutinin (NT).

Application

Avian Influenza Hemagglutinin 2 antibody can be used for the detection of the Hemagglutinin protein from the H5N1 strain of avian influenza A in ELISA. It will detect 10 ng of free peptide at 1 μg/mL.

Physical form

Supplied in PBS containing 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service