Skip to Content
Merck
All Photos(1)

Key Documents

SAB3500496

Sigma-Aldrich

Anti-SIRT5 antibody produced in chicken

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NAD-dependent deacetylase sirtuin-5, Anti-SIR2L5, Anti-Sirtuin 5

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
ELISA (i)
WB
clone:
polyclonal
species reactivity:
human, mouse
citations:
2
technique(s):
indirect ELISA: suitable
western blot: suitable

biological source

chicken

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

human, mouse

technique(s)

indirect ELISA: suitable
western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SIRT5(23408)

General description

SIRT5 (Sirtuin 5) is a mitochondrial matrix protein belonging to the class III of the sirtuin family. It is localized inside the mitochondria and within the cytosol and nucleus. The protein is widely expressed in the brain, heart, liver and kidney. It is composed of very short amino and carboxyl terminal sequences flanking region with the conserved, catalytic sirtuin center domain.

Immunogen

SIRT5 antibody was raised against a 17 amino acid peptide near the carboxy terminus of human SIRT5.

Application

Anti-SIRT5 antibody produced in chicken is suitable for western blot and indirect ELISA.

Biochem/physiol Actions

SIRT5 (Sirtuin 5) is associated with metabolism and aging process. It has been reported in a study that SIRT5 influences urea cycle pathway by NAD-dependent deacetylation and subsequent activation of carbamoyl phosphate synthetase 1 (CPS1), which plays a vital role in the initial step of urea cycle for the detoxification and removal of ammonia. In non-small cell-lung cancer cells. SIRT5 contributions have also been observed for the cancer cell growth and drug resistance. Study shows that SIRT5 may possess oncogenic property.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Linkage

The action of this antibody can be blocked using blocking peptide SBP3500496.

Physical form

Supplied in PBS with 0.02% sodium azide.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Nakagawa et al.
Cell, 137(3), 560-570 (2009-05-05)
Sirtuins are NAD-dependent protein deacetylases that connect metabolism and aging. In mammals, there are seven sirtuins (SIRT1-7), three of which are associated with mitochondria. Here, we show that SIRT5 localizes in the mitochondrial matrix and interacts with carbamoyl phosphate synthetase
Xiao-bin Lv et al.
Scientific reports, 5, 17940-17940 (2015-12-15)
SUN2, a key component of LINC (linker of nucleoskeleton and cytoskeleton) complex located at the inner nuclear membrane, plays unknown role in lung cancer. We found that SUN2 expression was decreased in lung cancer tissue compared with paired normal tissues

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service