Skip to Content
Merck
All Photos(2)

Documents

MTOX1010P24

Sigma-Aldrich

HepaRG 5F Control Cells

human female liver (Source Disease: Hepatocarcinoma and Hepatitis C)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352207
NACRES:
NA.24

product name

HepaRG 5F Control Cells, one assay ready, 24 well plate

biological source

human female liver (Source Disease: Hepatocarcinoma and Hepatitis C)

Quality Level

form

liquid

storage temp.

room temp

General description

HepaRG is a human hepatoma cell line. The cells possess a pseudodiploid karyotype and have been characterized as an oval ductular bipotent hepatic cell line as they have the ability to differentiate into both biliary and hepatocyte lineages in the presence of DMSO. HepaRG cells express the major xenobiotic sensors (PXR, CAR and AhR), drug transporters, phase I and II drug metabolizing enzymes as well as key hepatic transcription factors involved in stress response pathways.

Application

See technical bulletin for detailed protocols

Features and Benefits

Sigma′s HepaRG 5F Clone exhibits improved growth characteristics and enhanced functionality, expressing a large panel of drug metabolism enzymes; including cytochrome P450 enzymes CYP1A2, CYP2B6, CYP2C8/9, CYP2C19, CYP2D6, CYP2E1, and CYP3A4, nuclear receptors AhR, PXR, CAR, LXR, and FXR, and phase II enzymes UGT, SULT, NAT and GST.

  • The 24 well format provides delivery of ready-to-use liver cells for performing various experiments.

Quality

Tested for Mycoplasma, sterility, post-freeze viability, short terminal repeat (STR) analysis for cell line identification, cytochrome oxidase I (COI) analysis for cell line species confirmation

Legal Information

These products are covered by the License Agreement as described in Exhibit 1 and 2, in the technical bulletin.
HepaRG is a trademark of BioPredic International company

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service