HS120254
Wide mouth safety labelled wash bottle
capacity 500 mL, label, Methanol, PE-LD
All Photos(1)
About This Item
UNSPSC Code:
41103209
NACRES:
NB.43
material
PE-LD
green bottle
low-density polyethylene bottle
polypropylene cap
wide-mouth bottle
feature
label Methanol
vented
with cap
packaging
pack of 6 ea
manufacturer/tradename
Heathrow Scientific HS120254
capacity
500 mL
Related Categories
General description
Clear and informative product safety information.
The information provided on the front of each bottle is specific to the product contained and includes the following — the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information and Hazard Codes. You also have the option of using other liquids within the bottles and creating the appropriate safety information on the outside of the bottle by using the ‘Self Labeling′ versions. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
Self Labeling Bottles must only be used with chemicals that are safe and compatible with LDPE
The information provided on the front of each bottle is specific to the product contained and includes the following — the Chemical Abstract Service (CAS) No., National Fire Protection Association (NFPA) codes and symbols, Protective Equipment Symbol (PES), Target Organ Information and Hazard Codes. You also have the option of using other liquids within the bottles and creating the appropriate safety information on the outside of the bottle by using the ‘Self Labeling′ versions. The closure and stem are molded in one piece to make them leak-proof. The angled neck and tip provide a uniform stream of liquid and the tip can be cut back if required to increase the flow.
- Wide bottle mouth makes filling easier
- Non-metallic vent that prevents the risk of solvent leakage
- With the exception of the bottle designed for Sodium Hypochlorite, which is a white LDPE bottle for light protection and has a HDPE closure, the bottles are made from LDPE, with a PP closure
- Six named chemicals, a self labeling and assorted pack of bottles to choose from
- 500 mL bottle size
- Supplied in packs of 6
Self Labeling Bottles must only be used with chemicals that are safe and compatible with LDPE
Choose from one of the most recent versions:
Certificates of Analysis (COA)