Skip to Content
Merck
All Photos(2)

Key Documents

HPA019845

Sigma-Aldrich

Anti-SSH1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Protein phosphatase Slingshot homolog 1, Anti-SSH-1L, Anti-hSSH-1L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41
conjugate:
unconjugated
application:
IHC
clone:
polyclonal
species reactivity:
human
citations:
4
technique(s):
immunohistochemistry: 1:200- 1:500

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

LCEKDVKKKLEFGSPKGRSGSLLQVEETEREEGLGAGRWGQLPTQLDQNLLNSENLNNNSKRSCPNGMEDDAIFGILNKVKPSYKSCADCMYPTASGAPEASRERCEDPNAPAICTQPAFLPHITSSPVAHLASRS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SSH1(54434)

General description

SSH1 (Slingshot protein phosphatase 1) is a cofilin-phosphatase belonging to the slingshot (SSH). It is colocalized with F-actin on the cleavage furrow and midbody during cytokinesis.

Immunogen

Protein phosphatase Slingshot homolog 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

SSH1 (Slingshot protein phosphatase 1) is mainly involved in the actin filament organization. During cytokinesis, it reactivates ADF (actin-depolymerizing factor)/cofilin, which plays a major role in actin filament dynamics and cell migration. Apart from cofilin reactivation, it also dephosphorylates P-cofilin in vitro and in vivo. Overexpression of SSH1 has been reported in pancreatic cancer (PC) cells, which indicates its role in tumor cell migration via SSH1L/cofilin-1 signal pathway.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST73626

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Annie Cristhine Moraes Sousa-Squiavinato et al.
Cancer cell international, 21(1), 69-69 (2021-01-24)
Colorectal cancer (CRC) is among the deadliest cancers, wherein early dissemination of tumor cells, and consequently, metastasis formation, are the main causes of mortality and poor prognosis. Cofilin-1 (CFL-1) and its modulators, LIMK1/SSH1, play key roles in mediating the invasiveness
Noriko Kaji et al.
The Journal of biological chemistry, 278(35), 33450-33455 (2003-06-17)
During cytokinesis the actomyosin-based contractile ring is formed at the equator, constricted, and then disassembled prior to cell abscission. Cofilin stimulates actin filament disassembly and is implicated in the regulation of contractile ring dynamics. However, little is known about the
Ryusuke Niwa et al.
Cell, 108(2), 233-246 (2002-02-08)
The ADF (actin-depolymerizing factor)/cofilin family is a stimulus-responsive mediator of actin dynamics. In contrast to the mechanisms of inactivation of ADF/cofilin by kinases such as LIM-kinase 1 (LIMK1), much less is known about its reactivation through dephosphorylation. Here we report
Yufeng Wang et al.
Cancer letters, 360(2), 171-176 (2015-02-17)
Slingshot-1L (SSH1L), a cofilin-phosphatase, plays a role in actin dynamics and cell migration by reactivating cofilin-1. However, the expression of SSH1L in malignant diseases is poorly understood. The overexpression of SSH1L in cancerous tissue compared to the matched surrounding non-cancerous

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service