GE18-1035-32
CleanGel IEF precast gels
Polyacrylamide gel, 5% T/3% C, gel 110 mm L, pack of 5 ea
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41105300
NACRES:
NB.22
Recommended Products
material
polyester film
description
Polyacrylamide gel, 5% T/3% C
form
solid
feature
wetted part: no
packaging
pack of 5 ea
manufacturer/tradename
Cytiva 18103532
L × thickness
110 mm × 0.5 mm
color
Colorless
General description
CleanGel IEF precast gels are dried, "empty" gels, ready for reswelling in a carrier ampholyte solution of choice. CleanGel IEF gels offer improved gel performance with faster and more stable gradients, fewer artifacts, and easier zymogram assays. Before use, CleanGel IEF gels are rehydrated using the convenient GelPool accessory. Swelling returns the gel to its original thickness of 0.5 mm.
Features and Benefits
- Perform sensitive procedures using precast gels that are carefully washed and dried after casting to remove catalysts and unpolymerized material.
- Customize IEF separations by reswelling in a suitable mixture of carrier ampholytes and other additives such as detergents or denaturants.
- Handle mechanically stable, plastic-backed gels with confidence.
Storage and Stability
Please be aware this product may be shipped 90 days before the expiration date. For more information on the batch specific expiration date, please contact technical service.
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service