Skip to Content
Merck
All Photos(1)

Key Documents

D1010

Sigma-Aldrich

Diazald®-(N-methyl-14C)

~95% (Radiochemical Purity), solid

Synonym(s):

N-Methyl-N-nitroso-p-toluenesulfonamide

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.21

Assay

~95% (Radiochemical Purity)

Quality Level

form

solid

extent of labeling

20-60 mCi per mmol

storage temp.

2-8°C

Packaging

sealed ampule

Legal Information

Diazald is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service