Skip to Content
Merck
All Photos(1)

Documents

KC

90070550, insect embryo, Not specified

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

KC, 90070550

biological source

insect embryo

growth mode

Semi-adherent

karyotype

Not specified

morphology

Not specified

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Drosophila embryo

Cell Line Description

Resistant to osmolarity changes within 225-400 mosmols and to chloride levels 4-80mM; the usual Na/K ratio of 1.6 can be both increased and decreased considerably. Viruses present are non-pathogenic but can be induced when inoculated into a new host.

Application

Virus studies: rhabdovirus Drosophila X, picornavirus Drosophila C

Culture Medium

Schneider′s Drosophila medium + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Maintain cultures between 3-9x100,000 cells/ml. Cells may adhere to the tissue culture flask, shake gently to dislodge; 25°C; No CO2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service