Skip to Content
Merck
All Photos(1)

Documents

BAE-1

88031149, bovine cardiovascular , Endothelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

BAE-1, 88031149

biological source

bovine cardiovascular

growth mode

Adherent

karyotype

59 or 60

morphology

Endothelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Bovine aortic endothelium

Cell Line Description

Derived from the aorta of a Friesian cow. Cells form "sprouts" after confluence. Infected with Bovine Viral Diarrhoea Virus (BVDV) which is a common contaminant of ruminants and is non-hazardous to humans. Secretes proteoglycans and collagen.

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS).

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:4 i.e. seeding at 3-5x10,000 cells/cm2 using 0.25% trypsin; 5% CO2; 37°C. There is a tendancy for the cells to grow fibroblast-like until they are confluent. Once confluent the endothelial morphology prevail

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service