Skip to Content
Merck
All Photos(1)

Key Documents

HCA-7

6061902, human colon, Epithelial

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106514

product name

HCA-7, 06061902

biological source

human colon

growth mode

Adherent

karyotype

Not specified

morphology

Epithelial

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

relevant disease(s)

cancer

shipped in

dry ice

storage temp.

−196°C

Cell Line Origin

Human colon adenocarcinoma

Cell Line Description

Established from a colon adenocarcinoma of a 58 year-old female patient. The adenocarcinoma was moderately differentiated and determined to be Dukes′ stage B. Imperial College Innovations has stated that this cell line is not to be supplied to commercial organisations.

DNA Profile

STR-PCR Data: Amelogenin: X
CSF1PO: 11,14
D13S317: 9
D16S539: 10,11
D5S818: 9,12
D7S820: 11,12
THO1: 6,7
TPOX: 8,10
vWA: 14,16

Culture Medium

DMEM + 2mM Glutamine + 10% Foetal Bovine Serum (FBS)

Subculture Routine

Split sub-confluent cultures (70-80%) 1:2 to 1:10 using 0.05% trypsin/EDTA; 7.5% CO2; 37°C. Suggested seeding density 2-4x10,000 cells/cm2.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service