Skip to Content
Merck
All Photos(1)

Documents

AV38875

Sigma-Aldrich

Anti-ZFP36L2 antibody produced in rabbit

IgG fraction of antiserum

Synonym(s):

Anti-Zinc finger protein 36, C3H type-like 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

51 kDa

species reactivity

pig, human, rabbit, dog, rat, bovine, horse, mouse

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... ZFP36L2(678)

Related Categories

Immunogen

Synthetic peptide directed towards the N terminal region of human ZFP36L2

Biochem/physiol Actions

ZFP36L2 is a member of TIS11 family and contains a putative zinc finger domain with a repeating cys-his motif. It is an RNA-binding protein and binds to mRNA molecules that are highly expressed for self-renewal of erythroid progenitors and terminal erythroid differentiation. ZFP36L2 is required thymic development and prevention of malignant transformation of T cells.

Sequence

Synthetic peptide located within the following region: MSTTLLSAFYDVDFLCKTEKSLANLNLNNMLDKKAVGTPVAAAPSSGFAP

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lingbo Zhang et al.
Nature, 499(7456), 92-96 (2013-06-12)
Stem cells and progenitors in many lineages undergo self-renewing divisions, but the extracellular and intracellular proteins that regulate this process are largely unknown. Glucocorticoids stimulate red blood cell formation by promoting self-renewal of early burst-forming unit-erythroid (BFU-E) progenitors. Here we
Daniel J Hodson et al.
Nature immunology, 11(8), 717-724 (2010-07-14)
ZFP36L1 and ZFP36L2 are RNA-binding proteins (RBPs) that interact with AU-rich elements in the 3' untranslated region of mRNA, which leads to mRNA degradation and translational repression. Here we show that mice that lacked ZFP36L1 and ZFP36L2 during thymopoiesis developed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service