Skip to Content
Merck
All Photos(2)

Key Documents

AV100797

Sigma-Aldrich

Anti-SOX9 Antibody

rabbit polyclonal

Synonym(s):

Anti-SRY (sex determining region Y)-box 9 (campomelic dysplasia, autosomal sex-reversal)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
rabbit, goat, mouse, bovine, human, guinea pig, rat, dog
citations:
3
technique(s):
western blot: suitable

Product Name

Anti-SOX9 antibody produced in rabbit, IgG fraction of antiserum

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

IgG fraction of antiserum

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

56 kDa

species reactivity

rabbit, goat, mouse, bovine, human, guinea pig, rat, dog

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

NCBI accession no.

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SOX9(6662)

Immunogen

Synthetic peptide directed towards the N terminal region of human SOX9

Application

Anti-SOX9 antibody produced in rabbit is suitable for western blotting at a concentration of 2.5 μg/ml.

Biochem/physiol Actions

Sox9 is a transcription factor involved in a wide range of developmental processes. It is required for cartilage development, morphogenesis of face, testis differentiation and sex determination.

Sequence

Synthetic peptide located within the following region: PCPSGSGSDTENTRPQENTFPKGEPDLKKESEEDKFPVCIREAVSQVLKG

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Young-Hoon Lee et al.
Genesis (New York, N.Y. : 2000), 49(4), 200-208 (2011-02-11)
The Sox family of transcriptional regulators has been implicated in the control of a broad array of developmental processes. One member of this family SOX9 was first identified as a candidate gene for campomelic dysplasia (CD), a human syndrome affecting
S Morais da Silva et al.
Nature genetics, 14(1), 62-68 (1996-09-01)
Heterozygous mutations in SOX9 lead to a human dwarfism syndrome, Campomelic dysplasia. Consistent with a role in sex determination, we find that Sox9 expression closely follows differentiation of Sertoli cells in the mouse testis, in experimental sex reversal when fetal
W Bi et al.
Nature genetics, 22(1), 85-89 (1999-05-13)
Chondrogenesis results in the formation of cartilages, initial skeletal elements that can serve as templates for endochondral bone formation. Cartilage formation begins with the condensation of mesenchyme cells followed by their differentiation into chondrocytes. Although much is known about the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service