Skip to Content
Merck
All Photos(1)

Key Documents

NIST8554

IAEA-S-1 (Sulfur Isotopes in Silver Sulfide)

NIST®SRM®

Synonym(s):

Silver sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ag2S
CAS Number:
Molecular Weight:
247.80
MDL number:
UNSPSC Code:
41116107

isotopic purity

 34S

form

powder

packaging

pkg of 0.5 g

manufacturer/tradename

NIST®

mp

845 °C (lit.)

density

7.234 g/mL at 25 °C (lit.)

storage temp.

15-25°C

SMILES string

[Ag]S[Ag]

InChI

1S/2Ag.S

InChI key

OCWNMDUAUUAHSJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This Reference Material (RM) is an international measurement standard that defines the Vienna Cañon Diablo Troilite (VCDT) scale for relative differences in sulfur (S) isotope-number ratios.
SRM 8554_cert SRM 8554_SDS

Other Notes

Example analytes are listed below as a reference. Please download a current certificate at nist.gov/SRM for current analytes and certified values.
Sulfur -34

See certificate for values and more details at nist.gov/SRM.

Legal Information

NIST is a registered trademark of National Institute of Standards and Technology
SRM is a registered trademark of National Institute of Standards and Technology

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service