The diameter of these particles is not reported on a lot-to-lot basis. However, the expected size range is 1 - 2 mm in diameter by 5 mm in length. Please see the link below to review the NIST Certificate:
https://tsapps.nist.gov/srmext/certificates/720.pdf
NIST720
Sapphire heat capacity
NIST® SRM® 720
Synonym(s):
Synthetic Sapphire (α-Al2O3)
About This Item
Recommended Products
grade
certified reference material
Quality Level
form
solid
packaging
pkg of 15 g
manufacturer/tradename
NIST®
Related Categories
General description
SRM 720_cert
SRM 720_SDS
Application
Features and Benefits
- The variation of accuracy of enthalpy and heat capacity values as a function of temperature is presented in NIST certificate.
- Notes on disposal and transport are documented in MSDS.
Other Notes
Legal Information
Storage Class Code
13 - Non Combustible Solids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
-
What is the diameter of this material?
1 answer-
Helpful?
-
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service