Skip to Content
Merck
All Photos(1)

Key Documents

H0600000

Histamine dihydrochloride

European Pharmacopoeia (EP) Reference Standard

Synonym(s):

2-(4-Imidazolyl)ethylamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9N3 · 2HCl
CAS Number:
Molecular Weight:
184.07
Beilstein:
3624116
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

API family

histamine

manufacturer/tradename

EDQM

mp

249-252 °C (lit.)

application(s)

cleaning products
cosmetics
food and beverages
personal care
pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].NCCc1c[nH]cn1

InChI

1S/C5H9N3.2ClH/c6-2-1-5-3-7-4-8-5;;/h3-4H,1-2,6H2,(H,7,8);2*1H

InChI key

PPZMYIBUHIPZOS-UHFFFAOYSA-N

Gene Information

human ... HRH1(3269)

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Histamine dihydrochloride EP Reference standard, intended for use in laboratory tests only as specifically prescribed in the European Pharmacopoeia.

Packaging

The product is delivered as supplied by the issuing Pharmacopoeia. For the current unit quantity, please visit the EDQM reference substance catalogue.

Other Notes

Sales restrictions may apply.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yuki Bessho et al.
Neuroscience letters, 578, 106-110 (2014-07-06)
In mammals, it is established that histamine is a neurotransmitter and/or neuromodulator in the central nervous system. It is produced by the enzyme histidine decarboxylase (HDC) in the tuberomammillary nucleus of the posterior hypothalamus. However, HDC as well as histaminergic
Kevin B Freeman et al.
Psychopharmacology, 231(12), 2395-2403 (2014-01-09)
Recent research has demonstrated that the drug, histamine, can function as a punisher of cocaine self-administration. However, little is known about how drug punishers affect the maximum reinforcing effectiveness of drugs as reinforcers. The goal of the present study was
Guadalupe-Elide Morales-Figueroa et al.
ACS chemical neuroscience, 5(8), 637-645 (2014-06-03)
High levels of histamine H3 receptors (H3Rs) are found in the globus pallidus (GP), a neuronal nucleus in the basal ganglia involved in the control of motor behavior. By using rat GP isolated nerve terminals (synaptosomes), we studied whether H3R
Lydia M M Vermeer et al.
Experimental neurology, 263, 8-16 (2014-09-30)
Migraine is one of the most common neurological disorders, leading to more than 1% of total disability reported and over 68 million visits to emergency rooms or physician's offices each year in the United States. Three times as many women
Cui Cui et al.
Bone, 72, 43-52 (2014-12-03)
Serotonin (5-HT)--a monoamine with a variety of physiological functions--has recently emerged as a major regulator of bone mass. 5-HT is synthesized in the brain and the gut, and gut-derived 5-HT contributes to circulating 5-HT levels and is a negative modulator

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service