Skip to Content
Merck
All Photos(1)

Key Documents

938793

Sigma-Aldrich

Potassium phosphate tribasic ChemBeads

new

Synonym(s):

K3PO4, Tripotassium phosphate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
K3PO4
CAS Number:
Molecular Weight:
212.27
UNSPSC Code:
12352100

form

solid

Quality Level

composition

, 14-16 wt. % (loading of base)

SMILES string

[K].OP(O)(O)=O

InChI

1S/3K.H3O4P/c;;;1-5(2,3)4/h;;;(H3,1,2,3,4)/q3*+1;/p-3

InChI key

LWIHDJKSTIGBAC-UHFFFAOYSA-K

General description

Potassium phosphate tribasic exhibits high catalytic property in trans esterification reactions. It has bactericidal activity.

Application

Potassium phosphate tribasic has been used:
- in lysis buffer for the isolation of protein from Gram positive bacteria
- as a reagent for the scalable synthesis and post-modification of NU-1000 (mesoporous metal-organic framework)
- as a reagent for the plasma membrane preparation of Lilium davidii or Oryza sativa with high purity
ChemBeads are chemical coated glass beads. ChemBeads offer improved flowability and chemical uniformity perfect for automated solid dispensing and high-throughput experimentation. The method of creating ChemBeads uses no other chemicals or surfactants allowing the user to accurately dispense sub-milligram amounts of chemical.
For general uses, product is also available in powdered form (P5629)

related product

Product No.
Description
Pricing

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service