Skip to Content
Merck
All Photos(1)

Key Documents

66337

Supelco

Conductivity Standard

conductivity 300000 μS/cm at 25 °C

Synonym(s):

Conductivity Calibration Solution

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

analytical standard

Quality Level

shelf life

limited shelf life, expiry date on the label

concentration

10-25% KCl (T)

conductivity

297000-303000 μS/cm at 25 °C

density

1.063-1.135 g/cm3 at 20 °C

application(s)

cleaning products
cosmetics
environmental
flavors and fragrances
food and beverages
personal care

format

single component solution

Looking for similar products? Visit Product Comparison Guide

Application

Conductivity standard solution for calibration of conductivity sensors for conductivity measurements. It is delivered in a 500 mL package size.

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Carc. 2 - STOT RE 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Denis V Axenov-Gribanov et al.
Antonie van Leeuwenhoek, 109(10), 1307-1322 (2016-07-10)
Isolated ecosystems existing under specific environmental conditions have been shown to be promising sources of new strains of actinobacteria. The taiga forest of Baikal Siberia has not been well studied, and its actinobacterial population remains uncharacterized. The proximity between the
Liptak.GB and Venczel K
Analysis and Analyzers, Volume 2, 2 (2016)
Volunteer stream monitoring a methods manual null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service