327778
Polystyrene
analytical standard, for GPC, 90,000
Synonym(s):
PS
About This Item
Recommended Products
grade
analytical standard
for GPC
Quality Level
form
powder
mol wt
average Mw 90,000 (Typical)
technique(s)
gel permeation chromatography (GPC): suitable
format
neat
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service