Skip to Content
Merck
All Photos(1)

Key Documents

MABF1989

Sigma-Aldrich

Anti-MUC2 Antibody, clone CCP58

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
eCl@ss:
32160702
NACRES:
NA.41

antibody form

purified antibody

clone

CCP58
monoclonal

purified by

using protein G

species reactivity

human, mouse, human

concentration

(Please refer to lot specific datasheet.)

technique(s)

ELISA: suitable
immunohistochemistry: suitable
western blot: suitable

isotype

IgG1κ

UniProt accession no.

General description

KLH-conjugated linear peptide (MI-29) corresponding to 29 amino acids from human Mucin-2 containing one repeat unit of 23 amino acids and part of the next repeat of four amino acids.

Specificity

Clone CCP58 is a mouse monoclonal antibody that specifically detects Mucin-2 derived peptides, but does not react with Mucin-1 and 3 derived peptides.

Application

Western Blotting Analysis: A representative lot detected MUC2 in Western Blotting applications (Xing, P.X., et. al. (1992). J Natl Cancer Inst. 84(9):699-703).

Enzyme Immunoassay (ELISA) Analysis: A representative lot detected MUC2 in ELISA applications (Xing, P.X., et. al. (1992). J Natl Cancer Inst. 84(9):699-703).

Immunohistochemistry (Paraffin) Analysis: A representative lot detected MUC2 in Immunohistochemistry applications (Xing, P.X., et. al. (1992). J Natl Cancer Inst. 84(9):699-703).

Quality

Evaluated by Immunohistochemistry (Paraffin) in human small intestine tissue sections.

Immunohistochemistry (Paraffin) Analysis: A 1:250 dilution of this antibody detected Mucin-2 in human small intestine tissue sections.

Physical properties

540.30 kDa calculated.

Physical form

Purified mouse monoclonal antibody IgG1 in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Storage and Stability

Stable for 1 year at 2-8°C from date of receipt.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service