Skip to Content
Merck
All Photos(1)

Key Documents

MABE1930

Sigma-Aldrich

Anti-SIRT2 Antibody, clone 7G5

Synonym(s):

EC:2.3.1.286, NAD-dependent protein deacetylase sirtuin-2, NAD-dependent protein defatty-acylase sirtuin-2, Regulatory protein SIR2 homolog 2, SIR2-like protein 2

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
clone:
7G5, monoclonal
application:
IP
WB
species reactivity:
human
technique(s):
immunoprecipitation (IP): suitable
western blot: suitable
citations:

biological source

rat

Quality Level

antibody form

purified antibody

antibody product type

primary antibodies

clone

7G5, monoclonal

mol wt

calculated mol wt 43.18 kDa
observed mol wt ~40 kDa

purified by

using protein G

species reactivity

human

packaging

antibody small pack of 100 μg

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

isotype

IgG2a

epitope sequence

C-terminal half

Protein ID accession no.

UniProt accession no.

storage temp.

2-8°C

Gene Information

human ... SIRT2(22933)

Specificity

Clone 7G5 is a rat monoclonal antibody that detects SIRT2. It recognizes both the unphosphorylated SIRT2 as well as serine 331 phosphorylated SIRT2. It detects both isoforms 1 and 2.

Immunogen

A linear peptide corresponding to 14 amino acids surrounding phosphoserine 331 and 335 in human SIRT2.

Application

Quality Control Testing

Evaluated by Western Blotting in Human cerebral cortex tissue extracts.

Western Blotting Analysis: A 1:500 dilution of this antibody detected SIRT2 in Human cortex tissue extract.

Tested Applications

Western Blotting Analysis: A representative lot detected SIRT2 in Western Blotting application. (Pandithage, R., et al. (2008). J Cell Biol. 180(5):915-29).

Immunoprecipitation Analysis: A representative lot immunoprecipitated SIRT2 in Immunoprecipitation application (Pandithage, R., et al. (2008). J Cell Biol. 180(5):915-29).

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user.

Target description

NAD-dependent protein deacetylase sirtuin-2 (UniProt: Q8IXJ6; also known as EC:2.3.1.286, NAD-dependent protein defatty-acylase sirtuin-2, Regulatory protein SIR2 homolog 2, SIR2-like protein 2) is encoded by the SIRT2 (also known as SIR2L, SIR2L2) gene (Gene ID: 22933) in human. SIRT2 is a NAD-dependent protein deacetylase that deacetylates internal lysines on histone and -tubulin as well as several key transcription factors. Its deacetylase domain is localized in amino acids 57-338 and its nuclear export signal motif is in amino acids 41-51. SIRT2 participates in the modulation of multiple and diverse biological processes, including cell cycle control, genomic integrity, microtubule dynamics, cell differentiation, metabolic networks, and autophagy. It serves as an antephase checkpoint that prevents early mitotic entry in response to microtubule stress agents and control of cell cycle progression and genomic stability. It is reported to specifically deacetylate histone H4 at lysine 16 (H4K16ac) between the G2/M transition and metaphase enabling H4K20me1 deposition by KMT5A leading to ulterior levels of H4K20me2 and H4K20me3 deposition throughout cell cycle, and mitotic S-phase progression. SIRT2 is phosphorylated at serine 368 by CDK1/cyclin B at the G2/M transition and this phosphorylation regulates the delay in cell-cycle progression. However, phosphorylation at serine 368 by Cdk2/cyclin E complex inactivates SIRT2-mediated -tubulin deacetylation, and negatively regulates cell adhesion, cell migration, and neurite outgrowth during neuronal differentiation. Phosphorylation at serine 331 (isoforms 2 and 3) by Cdk2/cyclin E and Cdk2/cyclin A is reported to inhibit the catalytic activity of SIRT2. SIRT2 levels are up-regulated in response to low levels of glucose and anoxia-reoxygenation stress and down-regulated in response to high levels of glucose. It regulates blood glucose homeostasis by deacetylating and stabilizing phosphoenolpyruvate carboxykinase. (Ref.: Pandithage, R., et al. (2008). J. Cell Biol. 180(5); 915-929; North, BJ., and Verdin, E. (2007). J. Biol. Chem. 282(27); 19546-19555; North, BJ., et al. (2003). Mol. Cell. 11(2); 437-444; Dryden SC., et al. (2003). Mol. Cell Biol. 23(9); 173-185).

Physical form

Purified rat monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.

Reconstitution

1.0 mg/mL. Please refer to guidance on suggested starting dilutions and/or titers per application and sample type.

Storage and Stability

Recommended storage: +2°C to +8°C.

Other Notes

Concentration: Please refer to the Certificate of Analysis for the lot-specific concentration.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service