GF041
Monocyte Chemotactic Protein-1, Recombinant rat
The Macrophage/monocyte chemoattractant protein-1 (or MCP-1 protein) plays an important role in the inflammatory response of blood monocytes & tissue macrophages.
Synonym(s):
MCP-1
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
rat
Quality Level
Assay
>98% (SDS-PAGE and HPLC)
manufacturer/tradename
Chemicon®
technique(s)
cell culture | mammalian: suitable
input
sample type hematopoietic stem cell(s)
NCBI accession no.
UniProt accession no.
shipped in
dry ice
Gene Information
rat ... CCL2(24770)
General description
Macrophage/monocyte chemoattractant protein-1 (MCP-1) plays an important role in the inflammatory response of blood monocytes and tissue macrophages. Rat MCP-1 is a 14.1 kDa protein containing 125 amino acid residues.
Product Source: E.coli
Physical form
Lyophilized protein with no additives. The protein may appear as a haze or film which is difficult to see at the bottom of the vial.
Storage and Stability
The lyophilized powder, though stable at room temperature, is best stored desiccated at -20°C. After a quick spin, reconstitute in water to a concentration of 0.1-1.0 mg/mL. This solution can then be diluted into other aqueous buffers and stored at 4°C for one week or -20°C for future use. After reconstitution store in working aliquots at -20°C for up to 6 months.
Analysis Note
Specific Activity: Determined by its ability to chemoattract human monocytes using a concentration range of 10.0-100.0 ng/ml.
Legal Information
CHEMICON is a registered trademark of Merck KGaA, Darmstadt, Germany
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Lab on a chip, 22(12), 2352-2363 (2022-05-14)
Quantitative nucleic acid amplification tests (qNAATs) are critical in treating infectious diseases, such as in HIV viral load monitoring or SARS-CoV-2 testing, in which viral load indicates viral suppression or infectivity. Quantitative PCR is the gold standard tool for qNAATs;
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service